Dihydroergocristine methanesulphonate

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Dihydroergocristine mesylate

J60638.03
J60638.03EA 330 GBP
J60638.03
Dihydroergocristine methanesulphonate
Dihydroergocristine methanesulphonate

CAS No.: 24730-10-7 Molecular Formula: C36H45N5O8S Formula Weight: 707.80 Physical Form: Powder Appearance: White MDL No.: MFCD00153792


Applications: 5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

Formula: C₃₆H₄₅N₅O₈S
MW: 707.843 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153792
CAS Number: 24730-10-7
EINECS: 246-434-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR