Naltrexone hydrochloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

J60590.MD J60590.03
J60590.MDEA 137 GBP
J60590.MD J60590.03
Naltrexone hydrochloride
Naltrexone hydrochloride

CAS No.: 16676-29-2 Molecular Formula: C19H21NO4.HCl Formula Weight: 377.86 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Light Sensitive Appearance: White Solubility: Soluble in water at 50mg/ml Beilstein Reference: 3580333 MDL No.: MFCD00069324


Applications: A congener of naloxone and also an opioid antagonist

Formula: C₁₉H₂₁NO₄·HCl
MDL Number: MFCD00069324
CAS Number: 16676-29-2
EINECS: 240-723-0
Merck Index: 14,6363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR