Sodium-1-propanethiolate ≥95%, Technical Grade
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Purity: 95%
CAS No.: 6898-84-6
Molecular Formula: C3H7NaS
Formula Weight: 98.14
Physical Form: Solid
Sensitivity: Air and Moisture Sensitive
Appearance: White to pale gray
Odour: Stench
Beilstein Reference: 3556024
UN No.: UN3263
MDL No.: MFCD00043318
Formula:
C₃H₇NaS Storage Temperature: Ambient |
MDL Number:
MFCD00043318 CAS Number: 6898-84-6 EINECS: 230-005-5 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...