H-Glu-otbu 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-4-Amino-5-(tert-butoxy)-5-oxopentanoic acid, L-Glutamic acid 1-tert-butyl ester, 1-tert-Butyl L-Glutamate, H-Glu-OtBu, L-Glutamic acid 1-tert-Butyl ester

H65342.03 H65342.06
H65342.03EA 46.3 GBP
H65342.03 H65342.06
H-Glu-otbu 97%
H-Glu-otbu

Purity: 97% CAS No.: 45120-30-7 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 139-145° Beilstein Reference: 2414793 Optical Rotation: +13°(c=1 in water) MDL No.: MFCD00038562

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Melting Pt: 139…145 °C
MDL Number: MFCD00038562
CAS Number: 45120-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR