3-Bromo-2-fluorobenzaldehyde 97%
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Bromo-2-fluorobenzenecarbaldehyde
Purity: 97%
CAS No.: 149947-15-9
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₇H₄BrFO Melting Pt: 43…46 °C Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD09027089 CAS Number: 149947-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...