2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine 97+%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,3-Dihydro-7-azaindole
H64551.06
H64551.14
H64551.06EA
44.5
GBP
H64551.06
H64551.14
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine 97+%
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine
Purity: 97+%
CAS No.: 10592-27-5
Molecular Formula: C7H8N2
Formula Weight: 120.15
Physical Form: Powder
Appearance: White
Melting Point: 78-83°
Beilstein Reference: 116366
MDL No.: MFCD06659751
Formula:
C₇H₈N₂ Melting Pt: 78…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659751 CAS Number: 10592-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...