1-Boc-pyrrole-2-boronic acidpinacol ester 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1-carboxylate

H64171.03 H64171.06
H64171.03EA 63.8 GBP
H64171.03 H64171.06
1-Boc-pyrrole-2-boronic acidpinacol ester 98%
1-Boc-pyrrole-2-boronic acidpinacol ester

Purity: 98% CAS No.: 1072944-98-9 Molecular Formula: C15H24BNO4 Formula Weight: 293.17 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD11504960

Formula: C₁₅H₂₄BNO₄
MW: 293.17 g/mol
MDL Number: MFCD11504960
CAS Number: 1072944-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR