3-Amino-1-adamantanol ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Aminoadamantan-1-ol

H64137.14 H64137.06
H64137.14EA 123 GBP
H64137.14 H64137.06
3-Amino-1-adamantanol ≥98%
3-Amino-1-adamantanol

Purity: 98+% CAS No.: 702-82-9 Molecular Formula: C10H17NO Formula Weight: 167.25 Flash Point: 110°(230°F) Physical Form: Solid Melting Point: 265° lit. Beilstein Reference: 2802127 MDL No.: MFCD01821204

Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Melting Pt: 265 °C (lit.)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01821204
CAS Number: 702-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR