4-Bromo-2-methoxypyridine 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

H64026.03 H64026.06
H64026.03EA 35.9 GBP
H64026.03 H64026.06
4-Bromo-2-methoxypyridine 98%
4-Bromo-2-methoxypyridine

Purity: 98% CAS No.: 100367-39-3 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Storage Temperature: 2°C to 8°C Physical Form: Low melting solid Appearance: Off-white to pale yellow Melting Point: 25-26° Boiling Point: 87°/15mm Beilstein Reference: 115199 MDL No.: MFCD08277268

Formula: C₆H₆BrNO
Boiling Pt: 87 °C (15 mmHg)
Melting Pt: 25…26 °C
MDL Number: MFCD08277268
CAS Number: 100367-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR