Ethyl-O-Mesitylsulphonylacetohydroxamate ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
O-Mesitylsulphonylacetohydroxamic acid Ethyl Ester, Ethyl N-(mesitylsulphonyl)oxyacetimidate
H55470.03
H55470.06
H55470.03EA
22.8
GBP
H55470.03
H55470.06
Ethyl-O-Mesitylsulphonylacetohydroxamate ≥98%
Ethyl-O-Mesitylsulphonylacetohydroxamate
Purity: 98+%
CAS No.: 38202-27-6
Molecular Formula: C13H19NO4S
Formula Weight: 285.36
Storage Temperature: 2°C to 8°C
Melting Point: 54-56°
Beilstein Reference: 2144156
MDL No.: MFCD00009244
Formula:
C₁₃H₁₉NO₄S MW: 285.36 g/mol Melting Pt: 54…56 °C |
MDL Number:
MFCD00009244 CAS Number: 38202-27-6 EINECS: 253-825-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...