4-Chloro-N-(4-methoxyphenyl)benzamide 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

H55469.MD H55469.03
H55469.MDEA 60.1 GBP
H55469.MD H55469.03
4-Chloro-N-(4-methoxyphenyl)benzamide 97%
4-Chloro-N-(4-methoxyphenyl)benzamide

Purity: 97% CAS No.: 4018-82-0 Molecular Formula: C14H12ClNO2 Formula Weight: 261.71 MDL No.: MFCD00508611

Formula: C₁₄H₁₂ClNO₂
MW: 261.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00508611
CAS Number: 4018-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR