2-(trans-2-Cyanovinyl)benzeneboronic acid 95%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H52652.MD
H52652.03
H52652.MDEA
0
GBP
H52652.MD
H52652.03
2-(trans-2-Cyanovinyl)benzeneboronic acid 95%
2-(trans-2-Cyanovinyl)benzeneboronic acid
Purity: 95%
CAS No.: 850568-63-7
Molecular Formula: C9H8BNO2
Formula Weight: 172.98
MDL No.: MFCD06659825
Formula:
C₉H₈BNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06659825 CAS Number: 850568-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...