Hafnium(IV) isopropoxide isopropanol adduct 99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Hafnium(IV) i-propoxide monoisopropylate

H52288.14 H52288.22
H52288.14EA 305 GBP
H52288.14 H52288.22
Hafnium(IV) isopropoxide isopropanol adduct 99%
Hafnium(IV) isopropoxide isopropanol adduct

Purity: 99% CAS No.: 2171-99-5 Molecular Formula: C12H28HfO4.C3H8O Formula Weight: 474.94 UN No.: UN3181 MDL No.: MFCD00070460

Formula: C₁₂H₂₈HfO₄·C₃H₈O
MW: 474.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070460
CAS Number: 2171-99-5
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR