2-(Boc-amino)-5-pyridinemethanol 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: tert-Butyl (5-(hydroxymethyl)pyridin-2-yl)carbamate

H50077.03EA 166 GBP
H50077.03 H50077.06
2-(Boc-amino)-5-pyridinemethanol 97%
2-(Boc-amino)-5-pyridinemethanol
Formula: C₁₁H₁₆N₂O₃
MW: 224.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07186264
CAS Number: 169280-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR