3-(Di-tert-butylphosphonium)propane sulphonate 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Di-tert-butylphosphino)propane-1-sulphonic acid

H34253.MD H34253.03
H34253.MDEA 65.6 GBP
H34253.MD H34253.03
3-(Di-tert-butylphosphonium)propane sulphonate 97%
3-(Di-tert-butylphosphonium)propane sulphonate

Purity: 97% CAS No.: 1055888-89-5 Molecular Formula: C11H25O3PS Formula Weight: 268.35 MDL No.: MFCD12546062

Formula: C₁₁H₂₅O₃PS
MW: 268.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546062
CAS Number: 1055888-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR