6-Cyanobenzothiazole 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

H32454.MD H32454.03 H32454.06
H32454.MDEA 27.4 GBP
H32454.MD H32454.03 H32454.06
6-Cyanobenzothiazole 98%
6-Cyanobenzothiazole

Purity: 98% CAS No.: 58249-61-9 Molecular Formula: C8H4N2S Formula Weight: 160.20 Melting Point: 137-138° UN No.: UN3439 MDL No.: MFCD06659656

Formula: C₈H₄N₂S
Melting Pt: 137…138 °C
Storage Temperature: Ambient
MDL Number: MFCD06659656
CAS Number: 58249-61-9
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR