2,3,6-Trifluorobenzenesulphonyl chloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H32026.MD H32026.03
H32026.MDEA 33.7 GBP
H32026.MD H32026.03
2,3,6-Trifluorobenzenesulphonyl chloride
2,3,6-Trifluorobenzenesulphonyl chloride

Purity: 97%
CAS No.: 1017779-75-7
Molecular Formula: C6H2ClF3O2S
Formula Weight: 230.59
UN No.: UN3265
MDL No.: MFCD09832391

Formula: C₆H₂ClF₃O₂S
MDL Number: MFCD09832391
CAS Number: 1017779-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR