Dichloro(dimethylamino)phosphine 98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Dimethylphosphoramidous dichloride, (Dimethylamino)phosphonous Dichloride
H27856.14
H27856.06
H27856.14EA
232
GBP
H27856.14
H27856.06
Dichloro(dimethylamino)phosphine 98%
Dichloro(dimethylamino)phosphine
Purity: 98%
CAS No.: 683-85-2
Molecular Formula: C2H6Cl2NP
Formula Weight: 145.96
Storage Temperature: 2°C to 8°C
Flash Point: -23°(-9°F)
Boiling Point: 150°
UN No.: UN2924
MDL No.: MFCD00013613
Formula:
C₂H₆Cl₂NP Boiling Pt: 150 °C Density: 1.271 Flash Pt: –23 °C (–9 °F) |
MDL Number:
MFCD00013613 CAS Number: 683-85-2 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...