Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Bis(pentamethylcyclopentadienyl)cobalticinium hexafluorophosphate

H27761.03 H27761.06
H27761.03EA 75.7 GBP
H27761.03 H27761.06
Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate 98%
Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate

Purity: 98% CAS No.: 79973-42-5 Molecular Formula: C20H30CoF6P Formula Weight: 474.36 Melting Point: ca 305° dec. UN No.: UN3261 MDL No.: MFCD00269818

Formula: C₂₀H₃₀CoF₆P
MW: 474.35 g/mol
Melting Pt: ∼ 305 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00269818
CAS Number: 79973-42-5
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR