Di-tert-butyl-N,N-diethylphosphoramidite 94%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Di-tert-butyl diethylphosphoramidite

H27380.06 H27380.03
H27380.06EA 137 GBP
H27380.06 H27380.03
Di-tert-butyl-N,N-diethylphosphoramidite 94%
Di-tert-butyl-N,N-diethylphosphoramidite

Purity: 94% CAS No.: 117924-33-1 Molecular Formula: [(CH3)3CO]2PN(CH2CH3)2 Formula Weight: 249.33 Storage Temperature: 2°C to 8°C Flash Point: 78°(172°F) Refractive Index: 1.434 Boiling Point: 39-41°/0.3mm MDL No.: MFCD00134307

Formula: C₁₂H₂₈NO₂P
Boiling Pt: 39…41 °C (0.3 mmHg)
Density: 0.896
Flash Pt: 78 °C (172 °F)
MDL Number: MFCD00134307
CAS Number: 117924-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR