1,1'-Bis(dichlorophosphino)ferrocene 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

H27293.MD H27293.03
H27293.MDEA 36.4 GBP
H27293.MD H27293.03
1,1'-Bis(dichlorophosphino)ferrocene 98%
1,1'-Bis(dichlorophosphino)ferrocene

Purity: 98% CAS No.: 142691-70-1 Molecular Formula: C10H8Cl4FeP2 Formula Weight: 387.78 UN No.: UN3261 MDL No.: MFCD01630854

Formula: C₁₀H₈Cl₄FeP₂
Storage Temperature: Ambient
MDL Number: MFCD01630854
CAS Number: 142691-70-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR