2-(Diphenylphosphino)ethylamine 95%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H27070.03
H27070.09
H27070.03EA
154
GBP
H27070.03
H27070.09
2-(Diphenylphosphino)ethylamine 95%
2-(Diphenylphosphino)ethylamine
Purity: 95%
CAS No.: 4848-43-5
Molecular Formula: (C6H5)2PCH2CH2NH2
Formula Weight: 229.27
UN No.: UN2735
MDL No.: MFCD00233831
Formula:
(C₆H₅)₂PCH₂CH₂NH₂ Storage Temperature: Ambient |
MDL Number:
MFCD00233831 CAS Number: 4848-43-5 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...