Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine ≥98% may contain suspended dimethylamine hydrochloride crystals

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Bis(3,5-bis(trifluoromethyl)phenyl)chlorophosphane

H25883.06 H25883.03
H25883.06EA 227 GBP
H25883.06 H25883.03
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine ≥98% may contain suspended dimethylamine hydrochloride crystals
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine

Purity: 98+% CAS No.: 142421-57-6 Molecular Formula: C16H6ClF12P Formula Weight: 492.63 UN No.: UN3265 MDL No.: MFCD01630852

Formula: C₁₆H₆ClF₁₂P
MW: 492.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01630852
CAS Number: 142421-57-6
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR