Ethyl-3-thiopheneacetate 98%
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Purity: 98%
CAS No.: 37784-63-7
Molecular Formula: C8H10O2S
Formula Weight: 170.23
Flash Point: 103°(217°F)
Refractive Index: 1.5100
Boiling Point: 110-114°/6mm
Beilstein Reference: 122555
MDL No.: MFCD00005472
Formula:
C₈H₁₀O₂S Boiling Pt: 110…114 °C (6 mmHg) Density: 1.137 Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005472 CAS Number: 37784-63-7 EINECS: 253-665-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...