Methyl methylphenylphosphinate 98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
A18760.03
A18760.14
A18760.03EA
17.5
GBP
A18760.03
A18760.14
Methyl methylphenylphosphinate 98%
Methyl methylphenylphosphinate
Purity: 98%
CAS No.: 6389-79-3
Molecular Formula: C8H11O2P
Formula Weight: 170.15
Boiling Point: 148-149°/17mm
MDL No.: MFCD00013909
Formula:
C₈H₁₁O₂P Boiling Pt: 148…149 °C (17 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00013909 CAS Number: 6389-79-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...