Furil 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,2-di(2-furyl)ethane-1,2-dione, 1,2-Di(furan-2-yl)ethane-1,2-dione

A15386.06 A15386.14
A15386.06EA 31.5 GBP
A15386.06 A15386.14
Furil 98%
Furil

Purity: 98% CAS No.: 492-94-4 Molecular Formula: C10H6O4 Formula Weight: 190.15 Melting Point: 162-165° Beilstein Reference: 383882 MDL No.: MFCD00003241

Formula: C₁₀H₆O₄
MW: 190.15 g/mol
Melting Pt: 162…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00003241
CAS Number: 492-94-4
EINECS: 207-766-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR