2H-1,4-Benzothiazin-3(4H)-one ≥99%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2H-Benzo[b][1,4]thiazin-3(4H)-one
A11091.14
A11091.22
A11091.14EA
0
GBP
A11091.14
A11091.22
2H-1,4-Benzothiazin-3(4H)-one ≥99%
2H-1,4-Benzothiazin-3(4H)-one
Purity: 99%
CAS No.: 5325-20-2
Molecular Formula: C8H7NOS
Formula Weight: 165.22
Melting Point: 176-178°
Beilstein Reference: 131642
MDL No.: MFCD00006698
Formula:
C₈H₇NOS MW: 165.21 g/mol Melting Pt: 176…178 °C |
MDL Number:
MFCD00006698 CAS Number: 5325-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...