Methyl-4-(triphenylphosphonio)crotonate bromide 98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
A10443.06
A10443.09
A10443.06EA
38.9
GBP
A10443.06
A10443.09
Methyl-4-(triphenylphosphonio)crotonate bromide 98%
Methyl-4-(triphenylphosphonio)crotonate bromide
Purity: 98%
CAS No.: 2181-98-8
Molecular Formula: CH3O2CCH=CHCH2P(C6H5)3Br
Formula Weight: 441.32
Melting Point: ca 175° dec.
Beilstein Reference: 4173385
MDL No.: MFCD00031575
Formula:
CH₃O₂CCH=CHCH₂P(C₆H₅)₃Br MW: 441.32 g/mol Melting Pt: ∼ 175 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00031575 CAS Number: 2181-98-8 EINECS: 218-557-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...