Indium tin oxide ≥99.99% (metals basis) for vacuum deposition
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
ITO
42677.14
42677.22
42677.14EA
261
GBP
42677.14
42677.22
Indium tin oxide ≥99.99% (metals basis) for vacuum deposition
Indium tin oxide
Purity: 99.99%
CAS No.: 50926-11-9
Molecular Formula: In2O3:SnO2; 90:10 wt%
Physical Form: 3-12mm Pieces
MDL No.: MFCD00171662
Formula:
In₂O₃·(SnO₂)x Storage Temperature: Ambient |
MDL Number:
MFCD00171662 CAS Number: 50926-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...