Molybdenite naturally occurring mineral, grains ∼ 0.06-0.19 in
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
CAS No.: 1309-56-4
Molecular Formula: MoS2
Formula Weight: 160.07
Odour: Odourless
Solubility: Soluble in sulfuric and nitric acid
MDL No.: MFCD00003470
Applications: For grain mounts, XRD and microprobe standards, identification of unknown minerals
Formula:
MoS₂ Storage Temperature: Ambient |
MDL Number:
MFCD00003470 CAS Number: 1309-56-4 EINECS: 215-172-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...