N,N-Dimethylacrylamide ≥99.5% stabilised
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Stabilisation:
Stabilised with 4-methoxyphenol (MEHQ) 100 ppm
Stab. with 100ppm 4-methoxyphenol
Purity: 99.5%
CAS No.: 2680-03-7
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 71°(161°F)
Physical Form: Liquid
Density: 0.962
Refractive Index: 1.4730
Sensitivity: Light Sensitive
Appearance: Colourless
Boiling Point: 82-84°/21mm
Beilstein Reference: 1742219
UN No.: UN2810
MDL No.: MFCD00008626
Formula:
C₅H₉NO Boiling Pt: 82…84 °C (21 mmHg) Density: 0.962 Flash Pt: 71 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008626 CAS Number: 2680-03-7 EINECS: 220-237-5 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...