Aluminum silicon slug (99:1 wt%) ≥99.99% (metals basis), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42322.09
42322.18
42322.30
42322.09EA
33.9
GBP
42322.09
42322.18
42322.30
Aluminum silicon slug (99:1 wt%) ≥99.99% (metals basis), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Aluminum silicon slug
Purity: 99.99%
CAS No.: 11145-30-5
Molecular Formula: Al:Si; 99:1 wt%
Odour: Odourless
MDL No.: MFCD00143595
Storage Temperature:
Ambient |
MDL Number:
MFCD00143595 CAS Number: 11145-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...