Hydrogen tetranitratoaurate(III) hydrate (38,9 - 40,1% Au) ≥99.99% (metals basis), Premion®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

40431.03 40431.06
40431.03EA 132 GBP
40431.03 40431.06
Hydrogen tetranitratoaurate(III) hydrate (38,9 - 40,1% Au) ≥99.99% (metals basis), Premion®
Hydrogen tetranitratoaurate(III) hydrate

Decomposes in water. Soluble in HNO₃


Purity: 99.99% Molecular Formula: HAu(NO3)4.xH2O (x≈3) Formula Weight: 445.99(anhy) Physical Form: Crystalline Melting Point: ca 72° dec. Solubility: Decomposes in water. Soluble in HNO3 UN No.: UN1477 MDL No.: MFCD00046178

Formula: HAu(NO₃)₄·xH₂O(x≈3)
MW: 445.99 g/mol (anhy)
Melting Pt: ∼ 72 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00046178
UN: 1477
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR