Bis(diphenylphosphino)acetylene ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2-Bis(diphenylphosphino)ethyne

39277.02 39277.04
39277.02EA 54.3 GBP
39277.02 39277.04
Bis(diphenylphosphino)acetylene ≥97%
Bis(diphenylphosphino)acetylene

Purity: 97% CAS No.: 5112-95-8 Molecular Formula: (C6H5)2PC≡C(C6H5)2 Formula Weight: 394.39 Physical Form: Crystalline Melting Point: 85-87° MDL No.: MFCD00003045

Formula: C₂₆H₂₀P₂
MW: 394.394 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00003045
CAS Number: 5112-95-8
EINECS: 225-842-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR