Pentamethylcyclopentadienyliron dicarbonyl dimer

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

39266.03 39266.06
39266.03EA 108 GBP
39266.03 39266.06
Pentamethylcyclopentadienyliron dicarbonyl dimer
Pentamethylcyclopentadienyliron dicarbonyl dimer

CAS No.: 35344-11-7 Molecular Formula: C24H30Fe2O4 Formula Weight: 493.96 Physical Form: Crystalline Appearance: Red-purple UN No.: UN3466 MDL No.: MFCD00058862

Formula: C₂₄H₃₀Fe₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00058862
CAS Number: 35344-11-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR