Dicarbonylcyclopentadienylcobalt
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Cyclopentadienylcobalt dicarbonyl, Dicarbonylcyclopentadienylcobalt(I)
23136.03
23136.06
23136.03EA
51.2
GBP
23136.03
23136.06
Dicarbonylcyclopentadienylcobalt
Dicarbonylcyclopentadienylcobalt
CAS No.: 12078-25-0
Molecular Formula: C7H5CoO2
Formula Weight: 180.05
Flash Point: 26°(80°F)
Physical Form: Liquid
Sensitivity: Air Sensitive
Boiling Point: 139-140°/710mm
UN No.: UN3281
MDL No.: MFCD00009589
Applications: Catalyst for the cyclotrimerization of alkynes
Formula:
C₇H₅CoO₂ MW: 180.05 g/mol Boiling Pt: 139…140 °C (710 mmHg) Flash Pt: 26 °C (80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009589 CAS Number: 12078-25-0 EINECS: 235-139-8 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...