Safety shoes, lace-up, U-Manager, Venice
Supplier: U-Power
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MG20044/39 MG20044/40 MG20044/41 MG20044/42 MG20044/43 MG20044/44 MG20044/45 MG20044/46 MG20044/47 MG20044/38
UPOWMG20044/39EA 98.7 GBP
UPOWMG20044/39 UPOWMG20044/40 UPOWMG20044/41 UPOWMG20044/42 UPOWMG20044/43 UPOWMG20044/44 UPOWMG20044/45 UPOWMG20044/46 UPOWMG20044/47 UPOWMG20044/38
Safety shoes, lace-up, U-Manager, Venice
Shoes and Boots
Metal-free safety shoes made from soft full-grain leather and a slip-resistant, hard wearing PU sole.
  • Micro-Tech fibre
  • Executive plus anatomic and breathable insole with micro- cell for always cool and fresh feet
  • Natural fit width 11 for additional comfort
  • Airtoe protective Composite toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certifications: EN 20345:2011; S3