Slot blotters
Supplier: Cleaver Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CLEVCSL-S48EA 0 GBP
CLEVCSL-S48 CLEVCSL-S24
Slot blotters
Blotting Apparatus
Typical applications for this slot blotter include clone screening with DNA/RNA probes in Southern/Northern blots and immunological screening with antibodies in Western blots. The units incorporate precision lapped surfaces to ensure uniform blotting membrane contact and a leak proof gasket. These prevent lateral transfer of samples and smudging by ensuring that a complete vacuum is formed. Six thumbscrews ensure even and tight sealing for fast sample suck down.
  • Simple construction
  • Easy assembly
  • Alphanumeric sample identification