Utility knives, ReAkta
Supplier: THE SAFETY KNIFE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
REAKTAMSDBLUE REAKTASDBLACK REAKTAMHDBLUE REAKTAHDBLACK REAKAUTOHDRED
KNIFREAKTAMSDBLUEEA 11.3 GBP
KNIFREAKTAMSDBLUE KNIFREAKTASDBLACK KNIFREAKTAMHDBLUE KNIFREAKTAHDBLACK KNIFREAKAUTOHDRED
Utility knives, ReAkta
Knives
Carbon steel or stainless steel blade, glass filled nylon plastic body. The ReAkta utility knife has a unique mechanism where the blade locks in the 'out' position when a controlled cut is being made, however if the knife is dropped, shaken or jerked, the blade will automatically retract. Available in a heavy duty version with aluminium blade carrier and a standard duty with glass-filled nylon plastic blade carrier.
  • Aluminium or plastic blade carrier
  • Moulded in a tough glassed filled nylon plastic
  • Blade storage inside the knife
  • Security key for a controlled blade change
  • Reversible and replaceable blades
  • Easy blade change and cleaning procedure
  • Available as metal detectable version with stainless steel blade
The heavy duty 'auto retract' version of the ReAkta utility knife comes with an aluminium blade carrier. The blade will not lock in an 'out' position and will retract unless being used. Once you have punctured into the material, you may let go of the lever and the blade will stay out whilst cutting, once finished, the blade will automatically retract back in. The lock switch on the ReAkta allows the blade to be locked into the knife but if an attempt is made to lock the blade in the out position the blade will retract back into the knife.
All models are ideal for cutting plasterboard, carpet, lino, rubber, cable and cards.