Safety knives, Penguin 400
Supplier: THE SAFETY KNIFE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P400MBLUE P400CYELLOW P400CGREEN P400MRED P400MYELLOW P400MGREEN
KNIFP400MBLUEEA 28.1 GBP
KNIFP400MBLUE KNIFP400CYELLOW 233-0170 KNIFP400MRED KNIFP400MYELLOW KNIFP400MGREEN
Safety knives, Penguin 400
Knives
Carbon steel or stainless steel blade, PP body. The Penguin 400 is a one piece, lightweight, disposable safety knife with twin blade entry and handle.
  • No removable parts
  • Guarded blade entries
  • Handle grip
  • Also available as metal detectable version with stainless steel blade
Ideal for cutting stretch/bubble wrap, plastic bags, string and netting.