Avantor® ACE® CN-300, HPLC Columns, 3 µm

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ACE-214-1002EA 0 GBP
ACE-214-1002
Avantor® ACE® CN-300, HPLC Columns, 3 µm
Chromatography Columns
These Avantor® ACE® CN-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.

  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Capillary to preparative dimensions
  • Wide range of particle sizes (3, 5 and 10 µm)
  • Designed for peptide and protein separations
  • Method validation kits contain three different batches of the same phase - available on request

These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR