Safety knives, Fish 600
Supplier: THE SAFETY KNIFE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F600MYELLBLUE F600MREDBLUE F600MGREEN F600MBLUE F600CRED
KNIFF600MYELLBLUEEA 30.9 GBP
KNIFF600MYELLBLUE KNIFF600MREDBLUE KNIFF600MGREEN KNIFF600MBLUE 233-0169
Safety knives, Fish 600
Knives
Carbon steel or stainless steel blade, PP body.
  • No removable parts
  • Single piece moulding
  • Complete with hook blade
  • Disposable
  • Lanyard point
  • Also available as metal detectable version with stainless steel blade
The Fish 600 series is the first disposable version of the Fish design with hook blade. This lightweight, economical but very effective safety cutter is ideal for cutting strapping, bubble wrap, tape, cable ties, cardboard and netting.