Safety knives, Fish 200
Supplier: THE SAFETY KNIFE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F200BLUE F200YELLOW F200RED F200ORANGE F200MBLUEDETEC F200GREEN
KNIFF200BLUEEA 35.1 GBP
KNIFF200BLUE KNIFF200YELLOW KNIFF200RED 233-0168 233-0167 KNIFF200GREEN
Safety knives, Fish 200
Knives
Stainless steel blade, nylon plastic body. The Fish 200 is the original enclosed blade safety knife with hook blade and cover.
  • Textured surface for firm grip
  • Complete with hook blade
  • Removable hook blade cover
  • Reversible and replaceable blades
  • Lanyard point
  • Also available in metal detectable plastic
Suitable for cutting strapping, card, cable ties, bubble/shrink wrap, string and more.