Filtration fume cupboard, Circulaire®
Supplier: MONMOUTH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
K-CT1400 K-CT1800 K-CT1100 K-CT800
MONMK-CT1400EA 0 GBP
MONMK-CT1400 MONMK-CT1800 MONMK-CT1100 MONMK-CT800
Filtration fume cupboard, Circulaire®
Fume Hoods and Enclosures Ductless Fume Hoods
The Circulaire® filtration fume cabinets CT800 with Visionaire® touch screen control system utilise the carbon and HEPA filter technology. The deep bed carbon filters and the large working aperture of the cabinet offer unrivalled operator protection from the inhalation of fumes or particulates.
  • Carbon filter saturation alarm and low airflow alarm
  • Automatic face velocity air speed compensation
  • Visual display of actual air speed and carbon filter capacity
  • Low energy: cost saving and great ‘green’ credentials and low noise
  • Automatically recognised carbon and HEPA filter types/blends
  • ECO mode with PIR sensor – cabinet automatically saves energy when left unused for a pre-determined and adjustable period of time
  • Easy and safe change pre-filter system
  • Clear sides for maximum comfort and visibility and sloped front for easy viewing
Cabinet manufactured from zintec mild steel means years of corrosion free use.