Anti-Biotin Mouse Monoclonal Antibody (CF™ 770)
Supplier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20367-250UL 20367-50UL
20367-250ULEA 376 GBP
20367-250UL 20367-50UL
Anti-Biotin Mouse Monoclonal Antibody (CF™ 770)
Antibodies
Monoclonal mouse anti-biotin antibody reacts with biotin conjugated to antibodies or other proteins, and may show less non-specific binding to cells or tissues compared to streptavidin conjugates.