Anti-Vtg Rabbit Polyclonal Antibody [clone: CS-2]

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

170170-100 170170-500
CAYM170170-100EA 856 GBP
CAYM170170-100 CAYM170170-500
Anti-Vtg Rabbit Polyclonal Antibody [clone: CS-2]
Antibodies
Type: Primary
Antigen: Vtg
Clonality: Polyclonal
Clone: CS-2
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: common Carp, Flounder, Halibut, Tilapia, Turbot, Wrasse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR