Anti-Vtg Mouse Monoclonal Antibody [clone: JE-10D4]

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

170167-100 170167-500
CAYM170167-100EA 933 GBP
CAYM170167-100 CAYM170167-500
Anti-Vtg Mouse Monoclonal Antibody [clone: JE-10D4]
Antibodies
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: JE-10D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Carp, fathead Minnow, Roach, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR