Anti-Vtg Mouse Monoclonal Antibody [clone: ND-1C8]

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

170105-100 170105-500
CAYM170105-100EA 933 GBP
CAYM170105-100 CAYM170105-500
Anti-Vtg Mouse Monoclonal Antibody [clone: ND-1C8]
Antibodies
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: ND-1C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Atlantic Cod, killifish, largemouth Bass, Rainbow Trout, Sheepshead Minnow, striped Bass, Tilapia
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR