Anti-Ubiquitin Mouse Monoclonal Antibody [clone: 5B9-B3]

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

13722-50 13722-200
CAYM13722-50EA 773 GBP
CAYM13722-50 CAYM13722-200
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: 5B9-B3]
Antibodies
Type: Primary
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: 5B9-B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR