Human dermal blood endothelial cells (HDBEC)
Supplier: PROMOCELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C-12211 C-12225
C-12211EA 975 GBP
C-12211 C-12225
Human dermal blood endothelial cells (HDBEC)
Cells
Primary human dermal blood endothelial (HDBEC) cells are a sub-population of the human dermal endothelial cells. They are isolated from the dermis of juvenile foreskin and adult skin (different locations) from a single donor.
Blood endothelial cells have a key function in physiological processes like vessel tonus, capillary permeability, blood coagulation, fibrolysis and angiogenesis. It is also useful for disease studies of atherosclerosis, tumour genesis and thrombosis.
Order Now
SPECIFICATIONS
Environmentally Preferable